Methyl 4-amino-1,3-thiazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-5-(methoxycarbonyl)-1,3-thiazole
OR301121-1G
OR301121-5G
OR301121-250MG
APOSOR301121-1GEA
89.9
EUR
APOSOR301121-1G
APOSOR301121-5G
APOSOR301121-250MG
Methyl 4-amino-1,3-thiazole-5-carboxylate
Methyl 4-amino-1,3-thiazole-5-carboxylate
Formel:
C₅H₆N₂O₂S Molecular Weight: 158,178 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09033876 CAS-Nummer: 278183-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...