2-Methylquinoline-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-2-methyl-1-azanaphthalene, 3-Carboxy-2-methylquinoline

OR301188-100MG OR301188-1G
APOSOR301188-100MGEA 29 EUR
APOSOR301188-100MG APOSOR301188-1G
2-Methylquinoline-3-carboxylic acid 95%
2-Methylquinoline-3-carboxylic acid
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00510998
CAS-Nummer: 635-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR