2-Amino-3-bromo-5-iodopyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-iodopyridin-2-amine
OR301226-5G
APOSOR301226-5GEA
63.8
EUR
APOSOR301226-5G
2-Amino-3-bromo-5-iodopyridine 98%
2-Amino-3-bromo-5-iodopyridine
Formel:
C₅H₄BrIN₂ Molecular Weight: 298,907 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272065 CAS-Nummer: 697300-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...