4-Amino-5-methoxy-6-chloropyrimidine
Lieferant: Apollo Scientific
Synonyme:
4-Amino-6-chloro-5-methoxypyrimidine
OR302066-5G
APOSOR302066-5GEA
119
EUR
APOSOR302066-5G
4-Amino-5-methoxy-6-chloropyrimidine
4-Amino-5-methoxy-6-chloropyrimidine
Formel:
C₅H₆ClN₃O Molecular Weight: 159,574 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787635 CAS-Nummer: 5018-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...