1-Cyclopropyl-1H-imidazole 95%

Lieferant: Apollo Scientific

Synonyme: 1-Cyclopropylimidazole

OR302079-1G OR302079-250MG OR302079-5G
APOSOR302079-1GEA 348 EUR
APOSOR302079-1G APOSOR302079-250MG APOSOR302079-5G
1-Cyclopropyl-1H-imidazole 95%
1-Cyclopropyl-1H-imidazole
Formel: C₆H₈N₂
Molecular Weight: 108,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22690770
CAS-Nummer: 135207-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR