7-Methoxyquinoline-4-carboxylic acid
Lieferant: Apollo Scientific
OR302185-1G
OR302185-100MG
OR302185-250MG
APOSOR302185-1GEA
748
EUR
APOSOR302185-1G
APOSOR302185-100MG
APOSOR302185-250MG
7-Methoxyquinoline-4-carboxylic acid
7-Methoxyquinoline-4-carboxylic acid
Formel:
C₁₁H₉NO₃ Molecular Weight: 203,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11108759 CAS-Nummer: 816449-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...