4-Bromo-7-methoxyquinoline 95%

Lieferant: Apollo Scientific
Gefahr

OR302186-1G OR302186-5G
APOSOR302186-1GEA 186 EUR
APOSOR302186-1G APOSOR302186-5G
4-Bromo-7-methoxyquinoline 95%
4-Bromo-7-methoxyquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08063187
CAS-Nummer: 1070879-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR