4-Methyl-1-phenylquinolin-2(1H)-one

Lieferant: Apollo Scientific

OR302259-1G OR302259-250MG
APOSOR302259-1GEA 464 EUR
APOSOR302259-1G APOSOR302259-250MG
4-Methyl-1-phenylquinolin-2(1H)-one
4-Methyl-1-phenylquinolin-2(1H)-one
Formel: C₁₆H₁₃NO
Molecular Weight: 235,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00024050
CAS-Nummer: 2540-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR