5-(Pyridin-2-yl)-1H-pyrazol-4-amine
Lieferant: Apollo Scientific
Synonyme:
3-(Pyridin-2-yl)-1H-pyrazol-4-amine
OR302335-100MG
OR302335-250MG
OR302335-500MG
APOSOR302335-100MGEA
551
EUR
APOSOR302335-100MG
APOSOR302335-250MG
APOSOR302335-500MG
5-(Pyridin-2-yl)-1H-pyrazol-4-amine
5-(Pyridin-2-yl)-1H-pyrazol-4-amine
Formel:
C₈H₈N₄ Molecular Weight: 160,176 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22690777 CAS-Nummer: 896467-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...