5-Amino-1-methyl-1,2-dihydropyridin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-1-methylpyridin-2(1H)-one
OR302363-1G
OR302363-5G
APOSOR302363-1GEA
94.4
EUR
APOSOR302363-1G
APOSOR302363-5G
5-Amino-1-methyl-1,2-dihydropyridin-2-one
5-Amino-1-methyl-1,2-dihydropyridin-2-one
Formel:
C₆H₈N₂O Molecular Weight: 124,141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09814480 CAS-Nummer: 33630-96-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...