1-(2-Methoxyethyl)-1H-pyrazol-4-amine
Lieferant: Apollo Scientific
OR302397-100MG
OR302397-250MG
APOSOR302397-100MGEA
66.7
EUR
APOSOR302397-100MG
APOSOR302397-250MG
1-(2-Methoxyethyl)-1H-pyrazol-4-amine
1-(2-Methoxyethyl)-1H-pyrazol-4-amine
Formel:
C₆H₁₁N₃O Molecular Weight: 141,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09965625 CAS-Nummer: 948570-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...