5-Amino-3-methyl-4-nitroisoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-4-nitroisoxazol-5-amine, 3-Methyl-4-nitro-1,2-oxazol-5-amine

OR3024-10G OR3024-1G OR3024-5G
APOSOR3024-10GEA 654 EUR
APOSOR3024-10G APOSOR3024-1G APOSOR3024-5G
5-Amino-3-methyl-4-nitroisoxazole
5-Amino-3-methyl-4-nitroisoxazole
Formel: C₄H₅N₃O₃
Molecular Weight: 143,101 g/mol
Schmelzpunkt: 154…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067955
CAS-Nummer: 41230-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR