6-Aminopyridine-3-sulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(aminosulphonyl)pyridine, 2-Amino-5-sulphamoylpyridine

OR302537-1G OR302537-250MG
APOSOR302537-1GEA 273 EUR
APOSOR302537-1G APOSOR302537-250MG
6-Aminopyridine-3-sulphonamide
6-Aminopyridine-3-sulphonamide
Formel: C₅H₇N₃O₂S
Molecular Weight: 173,193 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07440120
CAS-Nummer: 57187-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR