2,3-Dihydro-1,4-benzodioxine-6-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR30258-1G OR30258-5G OR30258-10G
APOSOR30258-1GEA 21.9 EUR
APOSOR30258-1G APOSOR30258-5G APOSOR30258-10G
2,3-Dihydro-1,4-benzodioxine-6-carbonitrile
2,3-Dihydro-1,4-benzodioxine-6-carbonitrile
Formel: C₉H₇NO₂
Molecular Weight: 161,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00221462
CAS-Nummer: 19102-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR