7-Chloro-1-methyl-1H-pyrazolo[3,4-c]pyridine 97%

Lieferant: Apollo Scientific

OR302625-100MG OR302625-250MG
APOSOR302625-100MGEA 120 EUR
APOSOR302625-100MG APOSOR302625-250MG
7-Chloro-1-methyl-1H-pyrazolo[3,4-c]pyridine 97%
7-Chloro-1-methyl-1H-pyrazolo[3,4-c]pyridine
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265711
CAS-Nummer: 957760-15-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR