N-(1-Oxoindan-5-yl)acetamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2,3-Dihydro-1-oxo-1H-inden-5-yl)acetamide, 5-Acetamido-2,3-dihydro-1-oxo-1H-indene, 5-(Acetylamino)-2,3-dihydro-1-oxo-1H-indene
OR30266-1G
OR30266-25G
OR30266-5G
APOSOR30266-1GEA
69.6
EUR
APOSOR30266-1G
APOSOR30266-25G
APOSOR30266-5G
N-(1-Oxoindan-5-yl)acetamide 97%
N-(1-Oxoindan-5-yl)acetamide
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,211 g/mol Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00099465 CAS-Nummer: 58161-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...