6-Bromo-8-(2,6-dichlorophenyl)-9H-purine

Lieferant: Apollo Scientific
Achtung

OR302674-100MG OR302674-250MG
APOSOR302674-100MGEA 105 EUR
APOSOR302674-100MG APOSOR302674-250MG
6-Bromo-8-(2,6-dichlorophenyl)-9H-purine
6-Bromo-8-(2,6-dichlorophenyl)-9H-purine
Formel: C₁₁H₅BrCl₂N₄
Molecular Weight: 343,994 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22576475
CAS-Nummer: 1227958-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR