(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Lieferant: Apollo Scientific
OR302705-1G
OR302705-5G
APOSOR302705-1GEA
0
EUR
APOSOR302705-1G
APOSOR302705-5G
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Formel:
C₁₁H₁₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06202402 CAS-Nummer: 101555-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...