Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-(2,5-dimethylthiazol-4-yl)acetate

OR30301-1G OR30301-250MG
APOSOR30301-1GEA 102 EUR
APOSOR30301-1G APOSOR30301-250MG
Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate
Methyl (2,5-dimethyl-1,3-thiazol-4-yl)acetate
Formel: C₈H₁₁NO₂S
Molecular Weight: 185.243 g/mol
Siedepunkt: 66…68 °C (0.2 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02677686
CAS-Nummer: 306937-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR