6-Bromo-1H-indole-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR303143-1G
APOSOR303143-1GEA
299
EUR
APOSOR303143-1G
6-Bromo-1H-indole-2-carbonitrile
6-Bromo-1H-indole-2-carbonitrile
Formel:
C₉H₅BrN₂ Molecular Weight: 221,053 g/mol Schmelzpunkt: 190…192 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD22689155 CAS-Nummer: 1420537-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...