6-Bromo-3-methoxy-2-methylbenzonitrile
Lieferant: Apollo Scientific
OR303148-1G
OR303148-500MG
APOSOR303148-1GEA
538
EUR
APOSOR303148-1G
APOSOR303148-500MG
6-Bromo-3-methoxy-2-methylbenzonitrile
6-Bromo-3-methoxy-2-methylbenzonitrile
Formel:
C₉H₈BrNO Molecular Weight: 226,07 g/mol Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22689160 CAS-Nummer: 1420537-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...