Benzyl 4,5-bis(benzyloxy)pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR303156-1G
OR303156-500MG
APOSOR303156-1GEA
538
EUR
APOSOR303156-1G
APOSOR303156-500MG
Benzyl 4,5-bis(benzyloxy)pyridine-2-carboxylate
Benzyl 4,5-bis(benzyloxy)pyridine-2-carboxylate
Formel:
C₂₇H₂₃NO₄ Molecular Weight: 425,476 g/mol Schmelzpunkt: 117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD15071409 CAS-Nummer: 112334-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...