4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran

Lieferant: Apollo Scientific
Achtung

OR303176-1G OR303176-500MG
APOSOR303176-1GEA 671 EUR
APOSOR303176-1G APOSOR303176-500MG
4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran
4-Bromo-7-methoxy-2,2-dimethyl-2,3-dihydro-1-benzofuran
Formel: C₁₁H₁₃BrO₂
Molecular Weight: 257.124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22414513
CAS-Nummer: 185406-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR