tert-Butyl N-(2-methylphenyl)carbamate
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl o-tolylcarbamate
OR303188-1G
OR303188-500MG
OR303188-5G
APOSOR303188-1GEA
538
EUR
APOSOR303188-1G
APOSOR303188-500MG
APOSOR303188-5G
tert-Butyl N-(2-methylphenyl)carbamate
tert-Butyl N-(2-methylphenyl)carbamate
Formel:
C₁₂H₁₇NO₂ Molecular Weight: 207,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09031443 CAS-Nummer: 74965-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...