Ethyl [(5-amino-1,3,4-thiadiazol-2-yl)thio]acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl [(5-amino-1,3,4-thiadiazol-2-yl)sulphanyl]acetate, Ethyl 2-((5-amino-1,3,4-thiadiazol-2-yl)thio)acetate
OR30319-1G
OR30319-250MG
APOSOR30319-1GEA
415
EUR
APOSOR30319-1G
APOSOR30319-250MG
Ethyl [(5-amino-1,3,4-thiadiazol-2-yl)thio]acetate
Ethyl [(5-amino-1,3,4-thiadiazol-2-yl)thio]acetate
Formel:
C₆H₉N₃O₂S₂ Molecular Weight: 219,285 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173723 CAS-Nummer: 32418-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...