Methyl 6-nitrobenzo[b]thiophene-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Methoxycarbonyl)-6-nitro-1-benzothiophene, Methyl 6-nitro-1-benzothiophene-2-carboxylate
OR303196-1G
OR303196-5G
APOSOR303196-1GEA
0
EUR
APOSOR303196-1G
APOSOR303196-5G
Methyl 6-nitrobenzo[b]thiophene-2-carboxylate
Methyl 6-nitrobenzo[b]thiophene-2-carboxylate
Formel:
C₁₀H₇NO₄S Schmelzpunkt: 206…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040671 CAS-Nummer: 34084-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...