20-(tert-Butoxy)-20-oxoicosanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 19-carboxynonadecanoate, 20-(tert-Butoxy)-20-oxoarachidic acid
OR303212-1G
OR303212-5G
APOSOR303212-1GEA
26.1
EUR
APOSOR303212-1G
APOSOR303212-5G
20-(tert-Butoxy)-20-oxoicosanoic acid
20-(tert-Butoxy)-20-oxoicosanoic acid
Formel:
C₂₄H₄₆O₄ Molecular Weight: 398,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23136041 CAS-Nummer: 683239-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...