6-Amino-8-bromo-1,7-naphthyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Bromo-1,7-naphthyridin-6-amine

OR303217-1G OR303217-500MG
APOSOR303217-1GEA 538 EUR
APOSOR303217-1G APOSOR303217-500MG
6-Amino-8-bromo-1,7-naphthyridine
6-Amino-8-bromo-1,7-naphthyridine
Formel: C₈H₆BrN₃
Molecular Weight: 224,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702034
CAS-Nummer: 5912-35-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR