5-tert-Butyl-1,3-oxazole-4-carbohydrazide

Lieferant: Apollo Scientific

OR303222-1G OR303222-500MG
APOSOR303222-1GEA 538 EUR
APOSOR303222-1G APOSOR303222-500MG
5-tert-Butyl-1,3-oxazole-4-carbohydrazide
5-tert-Butyl-1,3-oxazole-4-carbohydrazide
Formel: C₈H₁₃N₃O₂
Molecular Weight: 183,208 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD23697496
CAS-Nummer: 1426904-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR