6-Amino-5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-5-bromopyrimidine-2,4(1H,3H)-dione

OR303234-1G OR303234-500MG
APOSOR303234-1GEA 567 EUR
APOSOR303234-1G APOSOR303234-500MG
6-Amino-5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
6-Amino-5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Formel: C₄H₄BrN₃O₂
Molecular Weight: 205.997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00830386
CAS-Nummer: 6312-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR