6-Bromo-7-ethoxy-1-methyl-1,2,3,4-tetrahydroquinolin-2-one
Lieferant: Apollo Scientific
OR303251-1G
OR303251-500MG
APOSOR303251-1GEA
538
EUR
APOSOR303251-1G
APOSOR303251-500MG
6-Bromo-7-ethoxy-1-methyl-1,2,3,4-tetrahydroquinolin-2-one
6-Bromo-7-ethoxy-1-methyl-1,2,3,4-tetrahydroquinolin-2-one
Formel:
C₁₂H₁₄BrNO₂ Molecular Weight: 284,149 g/mol Schmelzpunkt: 122…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD24038187 CAS-Nummer: 1392223-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...