Methyl 6-({[(tert-butoxy)carbonyl]amino}amino)-2,5-dichloropyrimidine-4-carboxylate

Lieferant: Apollo Scientific

OR303264-1G OR303264-500MG
APOSOR303264-1GEA 538 EUR
APOSOR303264-1G APOSOR303264-500MG
Methyl 6-({[(tert-butoxy)carbonyl]amino}amino)-2,5-dichloropyrimidine-4-carboxylate
Methyl 6-({[(tert-butoxy)carbonyl]amino}amino)-2,5-dichloropyrimidine-4-carboxylate
Formel: C₁₁H₁₄Cl₂N₄O₄
Molecular Weight: 337.159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24447579
CAS-Nummer: 1432892-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR