N'-[(1E)-(4-Amino-6-chloropyrimidin-5-yl)methylidene](tert-butoxy)carbohydrazide

Lieferant: Apollo Scientific

OR303265-1G OR303265-500MG
APOSOR303265-1GEA 538 EUR
APOSOR303265-1G APOSOR303265-500MG
N'-[(1E)-(4-Amino-6-chloropyrimidin-5-yl)methylidene](tert-butoxy)carbohydrazide
N'-[(1E)-(4-Amino-6-chloropyrimidin-5-yl)methylidene](tert-butoxy)carbohydrazide
Formel: C₁₀H₁₄ClN₅O₂
Molecular Weight: 271,703 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24447580
CAS-Nummer: 1432903-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR