6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1-methyl-3,4-dihydroquinolin-2(1H)-one, 6-Bromo-1-methyl-2-oxo-1,2,3,4-tetrahydroquinoline
OR303274-1G
OR303274-5G
APOSOR303274-1GEA
102
EUR
APOSOR303274-1G
APOSOR303274-5G
6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one
6-Bromo-3,4-dihydro-1-methylquinolin-2(1H)-one
Formel:
C₁₀H₁₀BrNO Molecular Weight: 240,097 g/mol Schmelzpunkt: 81…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18266837 CAS-Nummer: 1092523-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...