5-Acetyl-2,3-dichloropyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dichloro-5-ethanoylpyridine, 1-(5,6-Dichloropyridin-3-yl)ethanone, 1-(5,6-Dichloropyridin-3-yl)ethan-1-one
OR303284-1G
APOSOR303284-1GEA
69.6
EUR
APOSOR303284-1G
5-Acetyl-2,3-dichloropyridine 95%
5-Acetyl-2,3-dichloropyridine
Formel:
C₇H₅Cl₂NO Molecular Weight: 190,027 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697703 CAS-Nummer: 120800-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...