2,2,4,4,5,5,7,7-Octamethyl-3,6-dioxa-2,7-disilaoctane

Lieferant: Apollo Scientific

OR303302-1G OR303302-500MG
APOSOR303302-1GEA 180 EUR
APOSOR303302-1G APOSOR303302-500MG
2,2,4,4,5,5,7,7-Octamethyl-3,6-dioxa-2,7-disilaoctane
2,2,4,4,5,5,7,7-Octamethyl-3,6-dioxa-2,7-disilaoctane
Formel: C₁₂H₃₀O₂Si₂
Molecular Weight: 262,536 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190609
CAS-Nummer: 6730-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR