N-[(1S)-1-Phenylethyl]hydroxylamine oxalic acid

Lieferant: Apollo Scientific

OR303303-1G OR303303-500MG
APOSOR303303-1GEA 180 EUR
APOSOR303303-1G APOSOR303303-500MG
N-[(1S)-1-Phenylethyl]hydroxylamine oxalic acid
N-[(1S)-1-Phenylethyl]hydroxylamine oxalic acid
Formel: C₈H₁₁NO·C₂H₂O₄
Molecular Weight: 227,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12545903
CAS-Nummer: 78798-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR