N-[(1R)-1-Phenylethyl]hydroxylamine oxalic acid

Lieferant: Apollo Scientific

OR303306-1G OR303306-500MG
APOSOR303306-1GEA 180 EUR
APOSOR303306-1G APOSOR303306-500MG
N-[(1R)-1-Phenylethyl]hydroxylamine oxalic acid
N-[(1R)-1-Phenylethyl]hydroxylamine oxalic acid
Lagertemperatur: Raumtemperatur
MDL: MFCD11656048
CAS-Nummer: 118743-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR