5-Methyl-2-nitropyridin-3-ol

Lieferant: Apollo Scientific
Achtung

OR303307-1G OR303307-500MG
APOSOR303307-1GEA 671 EUR
APOSOR303307-1G APOSOR303307-500MG
5-Methyl-2-nitropyridin-3-ol
5-Methyl-2-nitropyridin-3-ol
Formel: C₆H₆N₂O₃
Molecular Weight: 154,123 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13181711
CAS-Nummer: 15128-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR