(3S)-3-Amino-2,2-dimethyl-3-phenylpropan-1-ol ≥ ee 99%
Lieferant: Apollo Scientific
OR303317-1G
OR303317-500MG
APOSOR303317-1GEA
1410
EUR
APOSOR303317-1G
APOSOR303317-500MG
(3S)-3-Amino-2,2-dimethyl-3-phenylpropan-1-ol ≥ ee 99%
(3S)-3-Amino-2,2-dimethyl-3-phenylpropan-1-ol
Formel:
C₁₁H₁₇NO Molecular Weight: 179,259 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22644810 CAS-Nummer: 156406-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...