Ethyl 2,6-dichloro-5-cyanopyridine-3-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR303340-1G OR303340-5G
APOSOR303340-1GEA 125 EUR
APOSOR303340-1G APOSOR303340-5G
Ethyl 2,6-dichloro-5-cyanopyridine-3-carboxylate 98%
Ethyl 2,6-dichloro-5-cyanopyridine-3-carboxylate
Formel: C₉H₆Cl₂N₂O₂
Molecular Weight: 245.062 g/mol
Schmelzpunkt: 67…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD22415252
CAS-Nummer: 919354-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR