3-Nitro-4-(pyridin-2-ylthio)cinammic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-{3-Nitro-4-[(pyridin-2-yl)thio]phenyl}prop-2-enoic acid, 3-{3-Nitro-4-[(pyridin-2-yl)thio]phenyl}acrylic acid

OR30337-1G OR30337-250MG OR30337-100MG
APOSOR30337-1GEA 63.8 EUR
APOSOR30337-1G APOSOR30337-250MG APOSOR30337-100MG
3-Nitro-4-(pyridin-2-ylthio)cinammic acid
3-Nitro-4-(pyridin-2-ylthio)cinammic acid
Formel: C₁₄H₁₀N₂O₄S
Molecular Weight: 302,305 g/mol
Schmelzpunkt: 177…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173720
CAS-Nummer: 175278-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR