Benzene-1,4-diylbis(dimethylsilanol)

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Phenylenebis(dimethylsilanol), 1,4-Bis[hydroxy(dimethyl)silyl]benzene

OR303375-5G
APOSOR303375-5GEA 75.4 EUR
APOSOR303375-5G
Benzene-1,4-diylbis(dimethylsilanol)
Benzene-1,4-diylbis(dimethylsilanol)
Formel: C₁₀H₁₈O₂Si₂
Molecular Weight: 226,42 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039516
CAS-Nummer: 2754-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR