2-{[(4-Methoxyphenyl)methyl]amino}-2-methylpropanenitrile
Lieferant: Apollo Scientific
OR303999-1G
OR303999-500MG
APOSOR303999-1GEA
336
EUR
APOSOR303999-1G
APOSOR303999-500MG
2-{[(4-Methoxyphenyl)methyl]amino}-2-methylpropanenitrile
2-{[(4-Methoxyphenyl)methyl]amino}-2-methylpropanenitrile
Formel:
C₁₂H₁₆N₂O Molecular Weight: 204,268 g/mol Schmelzpunkt: 67…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10014481 CAS-Nummer: 68220-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...