Pentan-3-one oxime

Lieferant: Apollo Scientific

OR30404-250MG OR30404-1G OR30404-5G
APOSOR30404-250MGEA 226 EUR
APOSOR30404-250MG APOSOR30404-1G APOSOR30404-5G
Pentan-3-one oxime
Pentan-3-one oxime
Formel: C₅H₁₁NO
Molecular Weight: 101,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00110982
CAS-Nummer: 1188-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR