(1-Methylazetidin-3-yl)methanol

Lieferant: Apollo Scientific

OR304317-1G OR304317-5G
APOSOR304317-1GEA 155 EUR
APOSOR304317-1G APOSOR304317-5G
(1-Methylazetidin-3-yl)methanol
(1-Methylazetidin-3-yl)methanol
Formel: C₅H₁₁NO
Molecular Weight: 101,147 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21661367
CAS-Nummer: 1499172-23-4
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR