2-Azetidinecarboxylic acid, 1-methyl-, (2S)-

Lieferant: Apollo Scientific

OR304321-1G OR304321-250MG OR304321-500MG
APOSOR304321-1GEA 1370 EUR
APOSOR304321-1G APOSOR304321-250MG APOSOR304321-500MG
2-Azetidinecarboxylic acid, 1-methyl-, (2S)-
2-Azetidinecarboxylic acid, 1-methyl-, (2S)-
Formel: C₅H₉NO₂
Molecular Weight: 115,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19229294
CAS-Nummer: 255882-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR