N-(3-Acetylphenyl)-N-methylacetamide 98%

Lieferant: Apollo Scientific
Achtung

OR30433-1G OR30433-500MG
APOSOR30433-1GEA 40.6 EUR
APOSOR30433-1G APOSOR30433-500MG
N-(3-Acetylphenyl)-N-methylacetamide 98%
N-(3-Acetylphenyl)-N-methylacetamide
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445602
CAS-Nummer: 325715-13-7
EINECS: 608-756-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR