N,N-Diethylazetidin-3-amine dihydrochloride

Lieferant: Apollo Scientific

OR304333-10G OR304333-1G OR304333-5G
APOSOR304333-10GEA 0 EUR
APOSOR304333-10G APOSOR304333-1G APOSOR304333-5G
N,N-Diethylazetidin-3-amine dihydrochloride
N,N-Diethylazetidin-3-amine dihydrochloride
Formel: C₇H₁₆N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18381080
CAS-Nummer: 149088-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR