5-Iodo-2-methyl-1,3-benzothiazole

Lieferant: Apollo Scientific

OR304339-1G OR304339-5G
APOSOR304339-1GEA 521 EUR
APOSOR304339-1G APOSOR304339-5G
5-Iodo-2-methyl-1,3-benzothiazole
5-Iodo-2-methyl-1,3-benzothiazole
Formel: C₈H₆INS
Molecular Weight: 275,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00643249
CAS-Nummer: 90414-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR